Invitrogen
Manufacturer Code:PA520811
Catalog # PIPA520811
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 15 amino acid peptide from near the amino terminus of human RIPK1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell death protein RIP; receptor (TNFRSF)-interacting serine-threonine kinase 1; Receptor-interacting protein 1; receptor-interacting protein kinase 1; Receptor-interacting serine/threonine-protein kinase 1; RIP; RIP RIP1 RIP-1 cell death protein RIP receptor interacting protein receptor-interacting protein 1 receptor-interacting serine/threonine-protein kinase 1 serine/threonine-protein kinase RIP; RIP-1; serine/threonine-protein kinase RIP
Gene Aliases: D330015H01Rik; Rinp; RIP; RIP-1; RIP1; RIPK1
UniProt ID: (Human) Q13546, (Mouse) Q60855
Entrez Gene ID: (Human) 8737, (Rat) 306886, (Mouse) 19766
Molecular Function: kinase non-receptor serine/threonine protein kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.