Proteintech
Manufacturer Code:145671AP150UL
Catalog # 50-172-9547
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RINT1 Fusion Protein Ag6145 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HsRINT 1 RAD50 interacting protein 1 RAD50 interactor 1 RINT 1 RINT1; HsRINT-1; RAD50 interactor 1; RAD50-interacting protein 1; RINT-1
Gene Aliases: 1500019C06Rik; 2810450M21Rik; RGD1560433; RINT-1; RINT1
UniProt ID: (Human) Q6NUQ1, (Mouse) Q8BZ36
Entrez Gene ID: (Human) 60561, (Mouse) 72772, (Rat) 296750
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.