Proteintech
Manufacturer Code:150371AP150UL
Catalog # 50-172-9922
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RING1 Fusion Protein Ag6763 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase RING1; Polycomb complex protein RING1; Polycomb complex protein RING1 ring finger protein 1 RING1 RING1A RNF1; Really interesting new gene 1 protein; RING finger protein 1; RING-type E3 ubiquitin transferase RING1; Transcription repressor Ring1A
Gene Aliases: RING1; RING1A; RNF1
UniProt ID: (Human) Q06587, (Rat) Q6MGB6, (Mouse) O35730
Entrez Gene ID: (Human) 6015, (Rat) 309626, (Mouse) 19763
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.