Novus Biologicals
Manufacturer Code:NBP266642
Catalog # NBP266642
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 27G2 |
Immunogen | Hybridoma produced by the fusion of splenocytes from mice immunized with full-length recombinant human Rin protein and mouse myeloma cells. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: Ras and Rab interactor 1; Ras and Rab interactor 1 ras inhibitor 1 Ras inhibitor JC99 ras inhibitor RIN1 Ras interaction/interference protein 1; ras inhibitor 1; Ras inhibitor JC99; ras inhibitor RIN1; Ras interaction/interference protein 1
Gene Aliases: RIN1
UniProt ID: (Human) Q13671
Entrez Gene ID: (Human) 9610
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.