Invitrogen
Manufacturer Code:PA5101812
Catalog # PIPA5101812
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human RIN1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ras and Rab interactor 1; ras and Rab interactor 1 ras inhibitor ras inhibitor 1 ras inhibitor JC99 ras inhibitor RIN1 ras interaction/interference protein 1 RIBA Rin1 RIT2 ROC2; ras inhibitor; ras inhibitor 1; Ras inhibitor JC99; ras inhibitor RIN1; Ras interaction/interference protein 1
Gene Aliases: RIN1
UniProt ID: (Human) Q13671, (Rat) P97680, (Mouse) Q921Q7
Entrez Gene ID: (Human) 9610, (Rat) 207119, (Mouse) 225870
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.