Proteintech
Manufacturer Code:245761AP150UL
Catalog # 50-173-4432
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RIM1 Fusion Protein Ag20161 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CORD7 KIAA0340 Rab 3 interacting molecule 1 Rab 3 interacting protein 2 RAB3IP2 RIM RIM 1 RIM1 RIMS1; Rab-3-interacting molecule 1; Rab-3-interacting protein 1; Rab-3-interacting protein 2; Rab3 effector; Rab3 interacting protein 1; rab3-interacting molecule 1; rab3-interacting protein 1; RAB3-interacting protein 2; regulating synaptic membrane exocytosis 1 beta; Regulating synaptic membrane exocytosis protein 1; RIM 1; Rim1b protein; synaptic exocytosis regulator 1
Gene Aliases: C030033M19Rik; CORD7; KIAA0340; mKIAA0340; Nbla00761; Rab3ip1; RAB3IP2; RIM; RIM1; RIM1a; RIM1alpha; RIMS1; Serg1
UniProt ID: (Human) Q86UR5, (Mouse) Q99NE5, (Rat) Q9JIR4
Entrez Gene ID: (Human) 22999, (Mouse) 116837, (Rat) 84556
Molecular Function:
G-protein modulator
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.