Invitrogen
Manufacturer Code:PA520276
Catalog # PIPA520276
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | human GST-tagged RIG-1 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antiviral innate immune response receptor RIG-I; DEAD (Asp-Glu-Ala-Asp) box polypeptide 58; DEAD box protein 58; DEAD-box protein 58; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide; DEAD/H box polypeptide RIG-I; Probable ATP-dependent RNA helicase DDX58; Retinoic acid-inducible gene 1 protein; Retinoic acid-inducible gene I protein; retinoic acid-inducible gene-I; RIG-1; RIG-I; RIG-I DEAD box protein 58 DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide RIG-1 RNA helicase RIG-I probable ATP-dependent RNA helicase DDX58 retinoic acid inducible gene I retinoic acid-inducible gene 1 protein retinoic acid-inducible gene I protein; RIG-I-like receptor 1; RLR-1; RNA helicase RIG-I
Gene Aliases: 6430573D20Rik; C330021E21; DDX58; RIG-I; RIGI; RLR-1; SGMRT2
UniProt ID: (Human) O95786, (Mouse) Q6Q899
Entrez Gene ID: (Human) 23586, (Mouse) 230073, (Rat) 297989
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.