Invitrogen
Manufacturer Code:PA557857
Catalog # PIPA557857
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human RIF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RAP1 interacting factor homolog; RAP1 interacting factor homolog yeast DKFZp434D1026 DKFZp781N1478 FLJ12870 rap1-interacting factor 1 homolog telomere-associated protein RIF1; Rap1-interacting factor 1 homolog; Telomere-associated protein RIF1
Gene Aliases: RIF1
UniProt ID: (Human) Q5UIP0
Entrez Gene ID: (Human) 55183
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.