Novus Biologicals
Manufacturer Code:NBP156877
Catalog # NBP156877
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to RIC8B(resistance to inhibitors of cholinesterase 8 homolog B (C. elegans)) The peptide sequence was selected from the middle region of RIC8B. Peptide sequence HQFRVMAAVLRHCLLIVGPTEDKTEELHSNAVNLLSNVPVSCLD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: brain synembrin; Brain synembrin hSyn Protein Ric-8B RIC8 RIC8B resistance to inhibitors of cholinesterase 8 homolog B (C. elegans) synembryn synembryn-B; Brain synembryn; hSyn; Protein Ric-8B; resistance to inhibitors of cholinesterase 8 homolog B; Synembryn-B
Gene Aliases: hSyn; RIC8; RIC8B
UniProt ID: (Human) Q9NVN3
Entrez Gene ID: (Human) 55188
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.