Novus Biologicals
Manufacturer Code:NBP273908AF647
Catalog # N273908A647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3G3 |
Immunogen | Full length recombinant protein of human RIC8A (NP_068751) produced in HEK293T cells. |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Protein Ric-8A; resistance to inhibitors of cholinesterase 8 homolog A; RIC8 RIC8A resistance to inhibitors of cholinesterase 8 homolog A (C. elegans) synembryn synembryn-A; Synembryn-A
Gene Aliases: RIC8; RIC8A
UniProt ID: (Human) Q9NPQ8
Entrez Gene ID: (Human) 60626
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.