Invitrogen
Manufacturer Code:PA518316
Catalog # PIPA518316
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (CLSSDPDSDPD) corresponding to the C-terminus amino acids of RIC8A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein Ric-8A; resistance to inhibitors of cholinesterase 8 homolog A; RIC8A synembryn resistance to inhibitors of cholinesterase 8 homolog A (C. elegans) MGC104517 MGC131931 RIC8 RIC8 protein resistance to inhibitors of cholinesterase 8 homolog A MGC148073 MGC148074; Synembryn-A
Gene Aliases: RIC8; RIC8A
UniProt ID: (Human) Q9NPQ8
Entrez Gene ID: (Human) 60626
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.