Invitrogen
Manufacturer Code:PA562436
Catalog # PIPA562436
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human RIC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: connexin 43-interacting protein 150 kDa; Connexin-43-interacting protein of 150 kDa; Guanine nucleotide exchange factor subunit RIC1; Protein RIC1 homolog; RAB6A GEF complex partner 1; RAB6A-GEF complex partner protein 1
Gene Aliases: bA207C16.1; CIP150; KIAA1432; RIC1
UniProt ID: (Human) Q4ADV7
Entrez Gene ID: (Human) 57589
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.