Proteintech
Manufacturer Code:106321AP150UL
Catalog # 50-172-6463
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RHOC Fusion Protein Ag0988 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aplysia ras-related homolog 9 (RhoC); ARH9 ARHC H9 Rho cDNA clone 9 RHOC RHOH9; h9; oncogene RHO H9; ras homolog 9 (RhoC); ras homolog gene family, member C; RAS-related homolog 9; Rho cDNA clone 9; Rho-related GTP-binding protein RhoC; rhoC GTPase; SIG-61; Silica-induced gene 61 protein; small GTP binding protein RhoC
Gene Aliases: AI324259; ARH9; ARHC; H9; RHOC; RHOH9
UniProt ID: (Human) P08134, (Mouse) Q62159
Entrez Gene ID: (Human) 389, (Mouse) 11853, (Rat) 295342
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.