Proteintech
Manufacturer Code:667331IG150UL
Catalog # 50-173-7145
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1B3D7 |
Immunogen | RHOA Fusion Protein Ag1141 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Aplysia ras-related homolog 12; ARH12 ARHA Rho cDNA clone 12 RHO12 RHOA RHOH12 Transforming protein RhoA; h12; oncogene RHO H12; Rho cDNA clone 12; small GTP binding protein RhoA; Transforming protein RhoA
Gene Aliases: ARH12; ARHA; RHO12; RHOA; RHOH12
UniProt ID: (Human) P61586
Entrez Gene ID: (Human) 387
Molecular Function:
G-protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.