Abnova
Manufacturer Code:MAB12503
Catalog # 89-934-856
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | A531 |
Immunogen | Native purified RHO from bovine retinal outer segments. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against native bovine RHO.
For Research Use Only
Protein Aliases: Rhodopsin; rhodopsin (opsin 2, rod pigment) (retinitis pigmentosa 4, autosomal dominant)
Gene Aliases: RHO
UniProt ID: (Bovine) P02699
Entrez Gene ID: (Bovine) 509933
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.