Invitrogen
Manufacturer Code:PA575922
Catalog # PIPA575922
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 168-213 of Human RHD. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: blood group antigen D; blood group protein RHD; Blood group Rh(D) polypeptide; Blood group Rh(D) polypeptide RHXIII Rh polypeptide 2 RhPII Rhesus D antigen CD240D RHD; CD240D; D antigen (DCS); Erythrocyte membrane glycoprotein Rh30; Rh blood group antigen Evans; Rh blood group protein; Rh blood group, D anitgen; Rh blood group, D antigen; RH polypeptide 2; Rh-like polypeptide; RhD blood group antigen; RhD polypeptide; Rhesus blood group CE and D; Rhesus blood group D antigen allele DIII type 7; Rhesus D antigen; Rhesus system D polypeptide; RhPII; RHXIII; truncated rhesus blood group D antigen; truncated rhesus D
Gene Aliases: CD240D; DIIIc; RH; RH30; Rh4; RHCED; RHD; RhDCw; RHDel; RHDVA(TT); RhII; RhK562-II; Rhl1; RhPI; RHPII; RHXIII
UniProt ID: (Human) Q02161, (Mouse) Q8CF94, (Rat) O88298
Entrez Gene ID: (Human) 6007, (Mouse) 19746, (Rat) 60414
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.