Novus Biologicals
Manufacturer Code:NBP2842480.1ML
Catalog # NB032176
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of RHD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: blood group antigen D; blood group protein RHD; Blood group Rh(D) polypeptide; blood group Rh(D) polypeptide CD240D DIIIc RH Rh blood group D antigen RH30 Rh4 RHCED RhDCw RHDel RHDVA(TT) RhII RhK562-II RhPI RHPII RHXIII; CD240D; D antigen (DCS); Rh blood group antigen Evans; Rh blood group, D anitgen; Rh blood group, D antigen; RH polypeptide 2; RhD blood group antigen; RhD polypeptide; Rhesus blood group D antigen allele DIII type 7; Rhesus D antigen; Rhesus system D polypeptide; RhPII; RHXIII; truncated rhesus blood group D antigen; truncated rhesus D
Gene Aliases: CD240D; DIIIc; RH; RH30; Rh4; RHCED; RHD; RhDCw; RHDel; RHDVA(TT); RhII; RhK562-II; RhPI; RHPII; RHXIII
UniProt ID: (Human) Q02161
Entrez Gene ID: (Human) 6007
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.