Abnova
Manufacturer Code:H00006007K
Catalog # 89-206-555
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human RHD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human RHD peptide using ARM Technology.
For Research Use Only
Protein Aliases: blood group antigen D; blood group protein RHD; Blood group Rh(D) polypeptide; CD240D; CD240D DIIIc MGC165007 RH RH30 RHCED RHDVA(TT) RHDel RHPII RHXIII Rh4 RhDCw RhII RhK562-II RhPI; D antigen (DCS); Rh blood group antigen Evans; Rh blood group, D anitgen; Rh blood group, D antigen; RH polypeptide 2; RhD blood group antigen; RhD polypeptide; Rhesus blood group D antigen allele DIII type 7; Rhesus D antigen; Rhesus system D polypeptide; RhPII; RHXIII; truncated rhesus blood group D antigen; truncated rhesus D
Gene Aliases: CD240D; DIIIc; RH; RH30; Rh4; RHCED; RHD; RhDCw; RHDel; RHDVA(TT); RhII; RhK562-II; RhPI; RHPII; RHXIII
UniProt ID: (Human) Q02161
Entrez Gene ID: (Human) 6007
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.