Invitrogen
Manufacturer Code:PA519383
Catalog # PIPA519383
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (ERQNSVYQSD) corresponding to the internal amino acids of RHCG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Ammonium transporter Rh type C; Rh family type C glycoprotein; Rh family, C glycoprotein; Rh glycoprotein kidney; Rh type C glycoprotein; RHCG Rh family C glycoprotein C15orf6 PDRC2 RHGK SLC42A3 Rh type C glycoprotein Rhesus blood group C glycoprotein; Rhesus blood group family type C glycoprotein; Rhesus blood group, C glycoprotein; Tumor-related protein DRC2
Gene Aliases: C15orf6; CDRC2; PDRC2; RHCG; RHGK; SLC42A3
UniProt ID: (Human) Q9UBD6
Entrez Gene ID: (Human) 51458
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.