Invitrogen
Manufacturer Code:PA543367
Catalog # PIPA543367
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human RHBG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ammonium transporter Rh type B; Rh family B glycoprotein (gene/pseudogene) SLC42A2; Rh family type B glycoprotein; Rh family, B glycoprotein; Rh type B glycoprotein; Rhesus blood group family type B glycoprotein; Rhesus blood group, B glycoprotein; Rhesus blood group-associated B glycoprotein
Gene Aliases: RHBG; SLC42A2
UniProt ID: (Bovine) Q95M77, (Dog) Q4VUI0, (Rabbit) Q95JD4, (Human) Q9H310, (Rat) Q68FT6, (Mouse) Q8BUX5
Entrez Gene ID: (Bovine) 282709, (Dog) 403538, (Rabbit) 100008876, (Human) 57127, (Horse) 100064158, (Guinea pig) 100723670, (Rat) 310625, (Mouse) 58176
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.