Invitrogen
Manufacturer Code:PA526978
Catalog # PIPA526978
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 423-450 amino acids from the C-terminal region of human RHBG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ammonium transporter Rh type B; Rh family type B glycoprotein; Rh family, B glycoprotein; Rh type B glycoprotein; Rhesus blood group family type B glycoprotein; Rhesus blood group, B glycoprotein; SLC42A2 Rh type B glycoprotein Rhesus blood group B glycoprotein ammonium transporter Rh type B
Gene Aliases: RHBG; SLC42A2
UniProt ID: (Human) Q9H310, (Mouse) Q8BUX5
Entrez Gene ID: (Human) 57127, (Mouse) 58176
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.