Invitrogen
Manufacturer Code:PA519369
Catalog # PIPA519369
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (PLRGGESDTR) corresponding to the C-terminus amino acids of RHBG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Ammonium transporter Rh type B; Rh family type B glycoprotein; Rh type B glycoprotein; RHBG Rh family B glycoprotein (gene/pseudogene) SLC42A2 Rh type B glycoprotein Rhesus blood group B glycoprotein; Rhesus blood group family type B glycoprotein
Gene Aliases: Rhbg
UniProt ID: (Mouse) Q8BUX5
Entrez Gene ID: (Mouse) 58176
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.