Novus Biologicals
Manufacturer Code:NBP29330820UL
Catalog # NB028502
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 206-315 of human RHBDD1 (NP_115652.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp547E052 EC 3.4.21 EC 3.4.21.- MGC117258 rhomboid domain containing 1 rhomboid domain-containing protein 1; Rhomboid domain-containing protein 1; Rhomboid-like protein 4; Rhomboid-related protein 4; RRP4
Gene Aliases: HSD-50; HSD50; RHBDD1; RHBDL4; RRP4
UniProt ID: (Human) Q8TEB9
Entrez Gene ID: (Human) 84236
Molecular Function: aspartic protease hydrolase protease transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.