Invitrogen
Manufacturer Code:PA5101836
Catalog # PIPA5101836
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human RGS7 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: regulator of G protein signaling 7 regulator of G-protein signaling 7 Regulator of G-protein signaling 7 (RGS7) regulator of G-protein signaling RGS7 regulator of G-protein signalling 7 Rgs7; Regulator of G-protein signaling 7; Regulator of G-protein signaling 7 (RGS7); regulator of G-protein signaling RGS7; regulator of G-protein signalling 7; RGS7
Gene Aliases: RGS7
UniProt ID: (Human) P49802, (Rat) P49803, (Mouse) O54829
Entrez Gene ID: (Human) 6000, (Rat) 54296, (Mouse) 24012
Molecular Function: G-protein modulator enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.