Novus Biologicals
Manufacturer Code:NBP200880
Catalog # NBP200880
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C1 |
Immunogen | Full length human recombinant protein of human RGS5 (NP_003608) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: MST092 MST106 MST129 MSTP032 MSTP092 MSTP106 MSTP129 regulator of G-protein signaling 5 regulator of G-protein signalling 5; Regulator of G-protein signaling 5; RGS5
Gene Aliases: MST092; MST106; MST129; MSTP032; MSTP092; MSTP106; MSTP129; RGS5
UniProt ID: (Human) O15539
Entrez Gene ID: (Human) 8490
Molecular Function: G-protein modulator enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.