Invitrogen
Manufacturer Code:PA540337
Catalog # PIPA540337
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human RGS4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Regulator of G-protein signaling 4; Regulator of G-protein signaling 4 MGC2124 MGC60244 RGP4 SCZD9; regulator of G-protein signalling 4; RGP4; RGS4; schizophrenia disorder 9
Gene Aliases: AA004315; AA597169; ESTM48; ESTM50; RGP4; RGS4; SCZD9
UniProt ID: (Rabbit) Q0R4E4, (Human) P49798, (Bovine) Q29RM9, (Mouse) O08899, (Rat) P49799
Entrez Gene ID: (Dog) 488666, (Rabbit) 100009014, (Horse) 100058342, (Sheep) 100381246, (Human) 5999, (Bovine) 617437, (Mouse) 19736, (Rat) 29480, (Guinea pig) 100717645
Molecular Function: G-protein modulator enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.