Invitrogen
Manufacturer Code:PA522332
Catalog # PIPA522332
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 205 of RGS4 (Uniprot IDNo.P49798) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Regulator of G-protein signaling 4; RGP4; RGS4; RP11-430G6.1 RGP4 SCZD9 schizophrenia disorder 9; schizophrenia disorder 9
Gene Aliases: AA004315; AA597169; ESTM48; ESTM50; RGP4; RGS4; SCZD9
UniProt ID: (Human) P49798, (Mouse) O08899, (Rat) P49799
Entrez Gene ID: (Human) 5999, (Mouse) 19736, (Rat) 29480
Molecular Function:
G-protein modulator
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.