Novus Biologicals
Manufacturer Code:NBP158901
Catalog # NBP158901
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C terminal of human RGS4 Peptide sequence EAQKKIFNLMEKDSYRRFLKSRFYLDLVNPSSCGAEKQKGAKSSADCASL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp761F1924 MGC2124 MGC60244 regulator of G-protein signaling 4 regulator of G-protein signalling 4 RGP4 schizophrenia disorder 9 SCZD9; Regulator of G-protein signaling 4; RGP4; schizophrenia disorder 9
Gene Aliases: RGP4; RGS4; SCZD9
UniProt ID: (Human) P49798
Entrez Gene ID: (Human) 5999
Molecular Function: G-protein modulator enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.