Novus Biologicals
Manufacturer Code:NBP29407120UL
Catalog # NB022703
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-100 of human RGS4 (NP_005604.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp761F1924 MGC2124 MGC60244 regulator of G-protein signaling 4 regulator of G-protein signalling 4 RGP4 schizophrenia disorder 9 SCZD9; Regulator of G-protein signaling 4; RGP4; schizophrenia disorder 9
Gene Aliases: RGP4; RGS4; SCZD9
UniProt ID: (Human) P49798
Entrez Gene ID: (Human) 5999
Molecular Function:
G-protein modulator
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.