Invitrogen
Manufacturer Code:PA519644
Catalog # PIPA519644
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide conjugated to KLH derived from within residues 150 to the C-terminus of Human RGS2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell growth-inhibiting gene 31 protein; cell growth-inhibiting protein 31; G0 to G1 switch regulatory 8, 24kD; G0/G1 switch regulatory protein 8; GIG31 G0S8 G0 to G1 switch regulatory 8 24kD G0/G1 switch regulatory protein 8 cell growth-inhibiting gene 31 protein cell growth-inhibiting protein 31 regulator of G-protein signaling 2; Regulator of G-protein signaling 2; regulator of G-protein signaling 2, 24kDa; regulator of G-protein signalling 2, 24kDa; RGS2
Gene Aliases: G0S8; GIG31; RGS2
UniProt ID: (Human) P41220
Entrez Gene ID: (Human) 5997, (Dog) 488584
Molecular Function:
G-protein modulator
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.