Novus Biologicals
Manufacturer Code:NB10056736
Catalog # NB10056736
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide within amino acid residues 1-50 of human RGS2 was used as immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell growth-inhibiting gene 31 protein; Cell growth-inhibiting gene 31 protein G0 to G1 switch regulatory 8 24kD G0/G1 switch regulatory protein 8 G0S8cell growth-inhibiting protein 31 regulator of G-protein signaling 2 regulator of G-protein signaling 2 24kDa regulator of G-protein signalling 2 24kD regulator of G-protein signalling 2 24kDa; cell growth-inhibiting protein 31; G0 to G1 switch regulatory 8, 24kD; G0/G1 switch regulatory protein 8; Regulator of G-protein signaling 2; regulator of G-protein signaling 2, 24kDa; RGS2
Gene Aliases: G0S8; GIG31; RGS2
UniProt ID: (Human) P41220
Entrez Gene ID: (Human) 5997
Molecular Function: G-protein modulator enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.