Invitrogen
Manufacturer Code:PA578498
Catalog # PIPA578498
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of mouse RGS14. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: rap1/rap2 interacting protein; RAP1/RAP2-interacting protein; Regulator of G-protein signaling 14; RGS14; RPIP1; RPIP1 regulator of G-protein signaling 14
Gene Aliases: Rgs14; RPIP1
UniProt ID: (Mouse) P97492, (Rat) O08773
Entrez Gene ID: (Mouse) 51791, (Rat) 114705
Molecular Function:
G-protein modulator
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.