Invitrogen
Manufacturer Code:PA5115841
Catalog # PIPA5115841
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human RGS14 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: rap1/rap2 interacting protein; rap1/rap2 interacting protein RAP1/RAP2-interacting protein Regulator of G-protein signaling 14 regulator of G-protein signalling 14 RGS14 RGSE RPIP1; RAP1/RAP2-interacting protein; Regulator of G-protein signaling 14; regulator of G-protein signalling 14; RGS14; RPIP1
Gene Aliases: RGS14; RPIP1
UniProt ID: (Human) O43566, (Mouse) P97492, (Rat) O08773
Entrez Gene ID: (Human) 10636, (Mouse) 51791, (Rat) 114705
Molecular Function:
G-protein modulator
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.