Novus Biologicals
Manufacturer Code:NBP158877
Catalog # NBP158877
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to RGS11(regulator of G-protein signaling 11) The peptide sequence was selected from the middle region of RGS11. Peptide sequence LRQPHRYVLDDAQLHIYMLMKKDSYPRFLKSDMYKALLAEAGIPLEMKRR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Regulator of G-protein signaling 11; regulator of G-protein signaling 11 regulator of G-protein signalling 11 RS11; regulator of G-protein signalling 11; RGS11
Gene Aliases: RGS11; RS11
UniProt ID: (Human) O94810
Entrez Gene ID: (Human) 8786
Molecular Function: G-protein modulator enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.