Invitrogen
Manufacturer Code:PA520519
Catalog # PIPA520519
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 17 amino acid peptide near the amino terminus of the human RGP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0258 retrograde Golgi transport protein RGP1 homolog; RAB6A-GEF complex partner protein 2; Retrograde Golgi transport protein RGP1 homolog; RGP1 retrograde golgi transport homolog; RGP1 retrograde golgi transport homolog (S. cerevisiae)
Gene Aliases: 1110029E03Rik; AI527196; F730001J03; KIAA0258; mKIAA0258; RGD1311757; RGP1
UniProt ID: (Human) Q92546, (Mouse) Q8BHT7
Entrez Gene ID: (Human) 9827, (Rat) 313493, (Mouse) 242406
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.