Novus Biologicals
Manufacturer Code:NBP198602
Catalog # NBP198602
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this antibody is RGP1 - C-terminal region. Peptide sequence FSVSLQTEERVQPEYQRRRGAGGVPSVSHVTHARHQESCLHTTRTSFSLP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RAB6A-GEF complex partner protein 2; Retrograde Golgi transport protein RGP1 homolog; retrograde Golgi transport protein RGP1 homolog RGP1 retrograde golgi transport homolog (S. cerevisiae); RGP1 retrograde golgi transport homolog
Gene Aliases: KIAA0258; RGP1
UniProt ID: (Human) Q92546
Entrez Gene ID: (Human) 9827
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.