Abnova
Manufacturer Code:PAB13031
Catalog # 89-116-284
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus 17 amino acids of human RGP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of RGP1.
For Research Use Only
Protein Aliases: KIAA0258; RAB6A-GEF complex partner protein 2; Retrograde Golgi transport protein RGP1 homolog; RGP1 retrograde golgi transport homolog
Gene Aliases: KIAA0258; RGP1
UniProt ID: (Human) Q92546
Entrez Gene ID: (Human) 9827
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.