Novus Biologicals
Manufacturer Code:NBP168976
Catalog # NBP168976
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to RGL3 (ral guanine nucleotide dissociation stimulator-like 3) The peptide sequence was selected from the middle region of RGL3. Peptide sequence RNFSSLRAILSALQSNPIYRLKRSWGAVSREPLSTFRKLSQIFSDENNHL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ00153 FLJ32585 FLJ44275 MGC126805 MGC138163 ral guanine nucleotide dissociation stimulator-like 3 RalGDS-like 3 RalGEF-like protein 3 mouse homolog; Ral guanine nucleotide dissociation stimulator-like 3; RalGDS-like 3; RalGEF-like protein 3, mouse homolog
Gene Aliases: RGL3
UniProt ID: (Human) Q3MIN7
Entrez Gene ID: (Human) 57139
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.