Invitrogen
Manufacturer Code:PA5110289
Catalog # PIPA5110289
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human RGL1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0959 mKIAA0959 Ral guanine nucleotide dissociation stimulator-like 1 RalGDS-like 1 Rgl Rgl1; Ral guanine nucleotide dissociation stimulator-like 1; RalGDS-like 1
Gene Aliases: KIAA0959; mKIAA0959; RGL; RGL1
UniProt ID: (Human) Q9NZL6, (Mouse) Q60695
Entrez Gene ID: (Human) 23179, (Mouse) 19731, (Rat) 289080
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.