Novus Biologicals
Manufacturer Code:NBP288144
Catalog # NB035766
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human RFX4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NYD-SP10 Regulatory factor X 4 regulatory factor X 4 (influences HLA class II expression) regulatory factor X4 Testis development protein NYD-SP10 transcription factor NYD-sp10 transcription factor RFX4; Regulatory factor X 4; regulatory factor X, 4 (influences HLA class II expression); Testis development protein NYD-SP10; Transcription factor RFX4; winged-helix transcription factor RFX4
Gene Aliases: NYD-SP10; RFX4
UniProt ID: (Human) Q33E94
Entrez Gene ID: (Human) 5992
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.