OriGene
Manufacturer Code:TA505916
Catalog # 50-167-6310
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3F7 |
Immunogen | Full length human recombit protein of human RFX3 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DNA binding protein RFX3; Regulatory factor X 3; regulatory factor X, 3 (influences HLA class II expression); regulatory factor X3; Transcription factor RFX3
Gene Aliases: RFX3
UniProt ID: (Human) P48380
Entrez Gene ID: (Human) 5991
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.