Invitrogen
Manufacturer Code:PA540355
Catalog # PIPA540355
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human RFX3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA binding protein RFX3; Regulatory factor X 3; regulatory factor X 3 (influences HLA class II expression); regulatory factor X, 3 (influences HLA class II expression); Transcription factor RFX3
Gene Aliases: C230093O12Rik; MRFX3; Rfx2; RFX3
UniProt ID: (Human) P48380, (Mouse) P48381
Entrez Gene ID: (Pig) 100523418, (Horse) 100051128, (Rabbit) 100009410, (Dog) 476339, (Human) 5991, (Mouse) 19726, (Rat) 361746
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.