Novus Biologicals
Manufacturer Code:NBP29464420UL
Catalog # NB023813
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 280-430 of human RFX3 (NP_602304.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA32F11.1 DNA binding protein RFX3 MGC87155 Regulatory factor X 3 regulatory factor X 3 (influences HLA class II expression) transcription factor RFX3; DNA binding protein RFX3; Regulatory factor X 3; regulatory factor X, 3 (influences HLA class II expression); Transcription factor RFX3
Gene Aliases: RFX3
UniProt ID: (Human) P48380
Entrez Gene ID: (Human) 5991
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.