Novus Biologicals
Manufacturer Code:NBP29424420UL
Catalog # NB023045
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 295-450 of human RFX2 (NP_000626.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA binding protein RFX2; DNA binding protein RFX2 DNA-binding protein RFX2 FLJ14226 HLA class II regulatory factor RFX2 Regulatory factor X 2 regulatory factor X 2 (influences HLA class II expression) trans-acting regulatory factor 2; DNA-binding protein RFX2; HLA class II regulatory factor RFX2; Regulatory factor X 2; regulatory factor X, 2 (influences HLA class II expression); trans-acting regulatory factor 2
Gene Aliases: RFX2
UniProt ID: (Human) P48378
Entrez Gene ID: (Human) 5990
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.