Invitrogen
Manufacturer Code:PA537032
Catalog # PIPA537032
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 475-520 of human Raftlin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell migration-inducing gene 2 protein; Cell migration-inducing gene 2 protein Raft-linking protein RFTN1 KIAA0084 ORF Names: MIG2; proliferation-inducing protein 10; proliferation-inducing protein 9; Raft-linking protein; Raftlin
Gene Aliases: 2310015N21Rik; AI853526; KIAA0084; MIG2; PIB10; PIG9; RAFTLIN; RFTN1; RGD1310777; RGD1563347
UniProt ID: (Human) Q14699, (Mouse) Q6A0D4
Entrez Gene ID: (Human) 23180, (Rat) 501095, (Mouse) 76438
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.