Novus Biologicals
Manufacturer Code:NBP29349120UL
Catalog # NB028859
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human RFTN1 (NP_055965.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell migration-inducing gene 2 protein; MIG2 PIB10 PIG9 RAFTLIN RFTN1 raftlin lipid raft linker 1; proliferation-inducing protein 10; proliferation-inducing protein 9; Raft-linking protein; Raftlin
Gene Aliases: KIAA0084; MIG2; PIB10; PIG9; RAFTLIN; RFTN1
UniProt ID: (Human) Q14699
Entrez Gene ID: (Human) 23180
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.