Novus Biologicals
Manufacturer Code:NBP300208R
Catalog # NB091657
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant RFK (Q969G6 Met1-His155). |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 0610038L10Rik; ATP:riboflavin 5'-phosphotransferase; ATP:riboflavin 5'-phosphotransferase EC 2.7.1.26 Flavokinase; Flavokinase; Riboflavin kinase
Gene Aliases: RFK; RIFK
UniProt ID: (Human) Q969G6
Entrez Gene ID: (Human) 55312
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.