Proteintech
Manufacturer Code:158131AP150UL
Catalog # 50-173-0501
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RFK Fusion Protein Ag8591 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 0610038L10Rik; ATP:riboflavin 5'-phosphotransferase; Flavokinase; Flavokinase RFK riboflavin kinase RIFK; KOI-4; Riboflavin kinase
Gene Aliases: 0610038L10Rik; AF031381; KOI-4; RFK; RIFK
UniProt ID: (Human) Q969G6, (Mouse) Q8CFV9
Entrez Gene ID: (Human) 55312, (Mouse) 54391, (Rat) 499328
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.