Invitrogen
Manufacturer Code:PA512418
Catalog # PIPA512418
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 170-200 amino acids from the central region of human RFC5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A1 36 kDa subunit; Activator 1 36 kDa subunit; Activator 1 subunit 5; replication factor C (activator 1) 5, 36.5kDa; Replication factor C 36 kDa subunit; Replication factor C subunit 5; Replication factor C subunit 5 Activator 1 subunit 5 Replication factor C 36 kDa subunit RF-C 36 kDa subunit RFC36 Activator 1 36 kDa subunit A1 36 kDa subunit RFC5; replication factor C, 36-kDa subunit; RF-C 36 kDa subunit; RFC, 36.5 kD subunit
Gene Aliases: RFC36; RFC5
UniProt ID: (Human) P40937
Entrez Gene ID: (Human) 5985, (Rat) 304528
Molecular Function: DNA binding protein DNA-directed DNA polymerase nucleic acid binding nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.