OriGene
Manufacturer Code:CF506183
Catalog # 50-166-4220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A8 |
Immunogen | Full length human recombit protein of human RFC4 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A1 37 kDa subunit; A1 RFC37; Activator 1 37 kDa subunit; Activator 1 subunit 4; replication factor C (activator 1) 4, 37kDa; Replication factor C 37 kDa subunit; Replication factor C subunit 4; RF-C 37 kDa subunit; RFC 37 kDa subunit
Gene Aliases: A1; AI894123; AU040575; RFC37; RFC4
UniProt ID: (Human) P35249, (Mouse) Q99J62
Entrez Gene ID: (Human) 5984, (Dog) 478667, (Mouse) 106344, (Rat) 288003
Molecular Function: DNA binding protein DNA-directed DNA polymerase nucleic acid binding nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.